Learn More
Eurofins Discoverx HePTP, 10 µg. (14-593)
Supplier:  Eurofins Discoverx 14593
Encompass
The HePTP product is purified N terminal 6xHis-tagged recombinant HePTP enzyme. This enzyme contains amino acids 22-end. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 10 μg of 1.629 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9019