Learn More
Sigma Aldrich Fine Chemicals Biosciences Heptyl acetate 98 fG1KG
Catalog No. 502898849
Encompass
Heptyl acetate is one of the main volatile constituents identified in Asian pear beer and Tosa-buntan essential oil.
Catalog No. 50-289-8849 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2547031KGK
Heptyl acetate is one of the main volatile constituents identified in Asian pear beer and Tosa-buntan essential oil.