Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptylphosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. H15355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15355G 5 g
H15351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15355G Supplier TCI America Supplier No. H15355G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4721-16-8
Molecular Formula C7H17O3P
Molecular Weight (g/mol) 180.184
MDL Number MFCD00015840
InChI Key VAJFLSRDMGNZJY-UHFFFAOYSA-N
PubChem CID 3870222
IUPAC Name heptylphosphonic acid
SMILES CCCCCCCP(=O)(O)O

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 180.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Heptylphosphonic Acid
EINECSNumber (2)-1995
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.