Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2064MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: HRB2/451. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: SKBR-3 Cells or Breast Cancers. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human HER2 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 185kDa.Chromosome: 17q12. Gene Symbol: ERBB2. Gene ID: 2064. Ensemble: ENSG00000141736. Uniprot: NP_001276865.
Catalog No. 50-188-4671