Learn More
Sino Biological Recombinant Human EphB2 Protein (GST Tag), Active
Catalog No. 502575083
Encompass_Preferred
Recombinant human EPHB2 (570-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5083 Supplier Sino Biological Supplier No. E2211G50
Recombinant human EPHB2 (570-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.