Learn More
HER2 anti-Human, Alexa Fluor™ 647, Clone: Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb), BD Phrmingen™
Shop All Waters Corporation ProductsDescription
Specifications
Specifications
| Antigen | HER2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb) |
| Conjugate | Alexa Fluor 647 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | CD340; ERBB2; HER-2; HER-2/neu; HER2; NEU; proto-oncogene c-ErbB-2 |
| Immunogen | A431 Cell Line Expressing EGFR |
| Purification Method | Affinity Chromatography |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.