Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER2 anti-Human, Alexa Fluor™ 647, Clone: Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb), BD Phrmingen™

Catalog No. p-200005185 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569979 100 Tests
BDB569980 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569979 Supplier BD Biosciences Supplier No. 569979
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biosimilar Monoclonal Antibody

HER2 Monoclonal antibody specifically detects HER2 in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HER2
Applications Flow Cytometry
Classification Monoclonal
Clone Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CD340; ERBB2; HER-2; HER-2/neu; HER2; NEU; proto-oncogene c-ErbB-2
Immunogen A431 Cell Line Expressing EGFR
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2064
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.