Learn More
Genscript Corporation Her2 Avi His Human
Catalog No. 5000057153
Encompass
ErbB2 also known as Neu and Her2 is a type I membrane glycoprotein belonging to the ErbB family of tyrosine kinase receptors It serves as a receptor for the epidermal growth factor (EGF) family of growth factors Upon activation ERBB2 triggers the MEMO1-RHOA-DIAPH1 signaling pathway leading to the phosphorylation and inhibition of GSK3B at the cell membrane
Catalog No. 50-000-57153 Supplier Genscript Corporation Supplier No. Z03806500
ErbB2 also known as Neu and Her2 is a type I membrane glycoprotein belonging to the ErbB family of tyrosine kinase receptors It serves as a receptor for the epidermal growth factor (EGF) family of growth factors Upon activation ERBB2 triggers the MEMO1-RHOA-DIAPH1 signaling pathway leading to the phosphorylation and inhibition of GSK3B at the cell membrane