Learn More
Medchemexpress LLC HER2/CD340 Human B 20ug
SDP
Catalog No. 5000182650
Encompass_Preferred
The HER2/CD340 protein is a dynamic tyrosine kinase that is essential in the neuregulin receptor complex and regulates microtubule dynamics Upon activation it triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B and promotes the association of APC and CLASP2 on the cell membrane to achieve microtubule stabilization HER2/CD340 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived HER2/CD340 protein expressed by HEK293 with C-Avi C-hFc labeled tag
Catalog No. 50-001-82650 Supplier Medchemexpress LLC Supplier No. HYP7237420UG
The HER2/CD340 protein is a dynamic tyrosine kinase that is essential in the neuregulin receptor complex and regulates microtubule dynamics Upon activation it triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B and promotes the association of APC and CLASP2 on the cell membrane to achieve microtubule stabilization HER2/CD340 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived HER2/CD340 protein expressed by HEK293 with C-Avi C-hFc labeled tag