Learn More
Medchemexpress LLC HER2/CD340 Human B 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP788825UG
Encompass_Preferred
The HER2/CD340 protein is a dynamic tyrosine kinase that is essential in the neuregulin receptor complex and regulates microtubule dynamics Upon activation it triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B and promotes the association of APC and CLASP2 on the cell membrane to achieve microtubule stabilization HER2/CD340 Protein Human (Biotinylated HEK293 His) is the recombinant human-derived Her2/CD340 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-95028