Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HER2/CD340 protein, human (HEK293, His) | >90.0% | 100UG
SDP

Catalog No. 5000175144
Encompass_Preferred

Recombinant human HER2 extracellular domain (residues T23-T652) expressed in HEK293 cells and purified with a C-terminal His tag. Supplied lyophilized for in vitro research applications, including binding assays, antibody characterization, and assay development.

  • Recombinant extracellular domain, 630 amino acids.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • C-terminal His tag for affinity purification and detection.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL).
  • Lyophilized formulation; refer to lot-specific COA for buffer.
  • Stable at -20°C unreconstituted; reconstituted stable 4°C for 1 week or -20°C/-80°C for longer.

Catalog No. 50-001-75144 Supplier Medchemexpress LLC Supplier No. HYP70254100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human HER2 extracellular domain (residues T23-T652) expressed in HEK293 cells and purified with a C-terminal His tag. Supplied lyophilized for in vitro research applications, including binding assays, antibody characterization, and assay development.

  • Recombinant extracellular domain, 630 amino acids.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • C-terminal His tag for affinity purification and detection.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL).
  • Lyophilized formulation; refer to lot-specific COA for buffer.
  • Stable at -20°C unreconstituted; reconstituted stable 4°C for 1 week or -20°C/-80°C for longer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.