Learn More
Medchemexpress LLC HER2/CD340 Human H 10ug
SDP
Catalog No. 5000180294
Encompass_Preferred
The HER2/CD340 protein is a dynamic tyrosine kinase that is essential in the neuregulin receptor complex and regulates microtubule dynamics Upon activation it triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B and promotes the association of APC and CLASP2 on the cell membrane to achieve microtubule stabilization HER2/CD340 Protein Human (HEK293) is the recombinant human-derived HER2/CD340 protein expressed by HEK293 with tag free
Catalog No. 50-001-80294 Supplier Medchemexpress LLC Supplier No. HYP7309410UG
The HER2/CD340 protein is a dynamic tyrosine kinase that is essential in the neuregulin receptor complex and regulates microtubule dynamics Upon activation it triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B and promotes the association of APC and CLASP2 on the cell membrane to achieve microtubule stabilization HER2/CD340 Protein Human (HEK293) is the recombinant human-derived HER2/CD340 protein expressed by HEK293 with tag free