Learn More
Medchemexpress LLC HER2/CD340 Human S 5ug | 5ug
SDP
Catalog No. 5000194240
Encompass_Preferred
The HER2/CD340 protein is a dynamic tyrosine kinase that is essential in the neuregulin receptor complex and regulates microtubule dynamics Upon activation it triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B and promotes the association of APC and CLASP2 on the cell membrane to achieve microtubule stabilization HER2/CD340 Protein Human (S310Y HEK293 His) is the recombinant human-derived HER2/CD340 protein expressed by HEK293 with C-10 His labeled tag and S310Y mutation
Catalog No. 50-001-94240 Supplier Medchemexpress LLC Supplier No. HYP786805UG
The HER2/CD340 protein is a dynamic tyrosine kinase that is essential in the neuregulin receptor complex and regulates microtubule dynamics Upon activation it triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B and promotes the association of APC and CLASP2 on the cell membrane to achieve microtubule stabilization HER2/CD340 Protein Human (S310Y HEK293 His) is the recombinant human-derived HER2/CD340 protein expressed by HEK293 with C-10 His labeled tag and S310Y mutation