Learn More
Medchemexpress LLC HER2/CD340 Mouse H 10ug
SDP
Catalog No. 5000179275
Encompass_Preferred
The HER2/CD340 protein is a multifunctional tyrosine kinase that is a component of the neuregulin receptor complex and regulates microtubule dynamics Activated HER2/CD340 triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B on the cell membrane and promotes the binding of APC and CLASP2 which is critical for microtubule stability HER2/CD340 Protein Mouse (HEK293 N-His) is the recombinant mouse-derived HER2/CD340 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-79275 Supplier Medchemexpress LLC Supplier No. HYP73929A10UG
The HER2/CD340 protein is a multifunctional tyrosine kinase that is a component of the neuregulin receptor complex and regulates microtubule dynamics Activated HER2/CD340 triggers the MEMO1-RHOA-DIAPH1 pathway inhibits GSK3B on the cell membrane and promotes the binding of APC and CLASP2 which is critical for microtubule stability HER2/CD340 Protein Mouse (HEK293 N-His) is the recombinant mouse-derived HER2/CD340 protein expressed by HEK293 with N-His labeled tag