Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Her2/CD340 protein, canine (HEK293, His) | >95.0% | 100 UG
SDP

Catalog No. 5000293030
Encompass_Preferred

HER2/CD340 Protein, Canine (HEK293, His) is a recombinant, partially truncated canine ErbB2 extracellular domain produced in HEK293 mammalian cells and supplied as a C-terminal His-tagged, lyophilized protein for research applications. It is suitable for biochemical and cell-based assays, including binding and functional activity measurements.

  • Recombinant partial extracellular domain (T23-T652) expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Supplied lyophilized for extended stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg), suitable for sensitive assays.
  • Store at -20°C; after reconstitution stable 4°C for 1 week or -20°C for long-term storage.
  • Demonstrated biological activity with ED50 0.4594 μg/mL in the presence of 0.6 μg/mL trastuzumab.

Catalog No. 50-002-93030 Supplier Medchemexpress LLC Supplier No. HYP74897100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HER2/CD340 Protein, Canine (HEK293, His) is a recombinant, partially truncated canine ErbB2 extracellular domain produced in HEK293 mammalian cells and supplied as a C-terminal His-tagged, lyophilized protein for research applications. It is suitable for biochemical and cell-based assays, including binding and functional activity measurements.

  • Recombinant partial extracellular domain (T23-T652) expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Supplied lyophilized for extended stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg), suitable for sensitive assays.
  • Store at -20°C; after reconstitution stable 4°C for 1 week or -20°C for long-term storage.
  • Demonstrated biological activity with ED50 0.4594 μg/mL in the presence of 0.6 μg/mL trastuzumab.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.