Learn More
Medchemexpress LLC HER2/CD340 protein, rat (HEK293, His) | >95.0% | 10 UG
SDP
Catalog No. 5000291696
Encompass_Preferred
Recombinant rat HER2/CD340 extracellular protein produced in HEK293 cells with a C-terminal 10xHis tag. Supplied lyophilized with supporting documentation (datasheet, COA, SDS), it is intended for research use in biochemical assays, binding studies, and assay development.
  • Expressed in HEK293 cells for mammalian glycosylation
  • C-terminal 10xHis tag for affinity purification and detection
  • Approximately 91 kDa molecular weight
  • Purity >95% as determined by reducing SDS-PAGE
  • Lyophilized formulation for convenient storage and transport; reconstitute per datasheet
  • Available in small microgram-scale sizes suitable for assay development
Catalog No. 50-002-91696 Supplier Medchemexpress LLC Supplier No. HYP7489510UG
Recombinant rat HER2/CD340 extracellular protein produced in HEK293 cells with a C-terminal 10xHis tag. Supplied lyophilized with supporting documentation (datasheet, COA, SDS), it is intended for research use in biochemical assays, binding studies, and assay development.
  • Expressed in HEK293 cells for mammalian glycosylation
  • C-terminal 10xHis tag for affinity purification and detection
  • Approximately 91 kDa molecular weight
  • Purity >95% as determined by reducing SDS-PAGE
  • Lyophilized formulation for convenient storage and transport; reconstitute per datasheet
  • Available in small microgram-scale sizes suitable for assay development