Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HER2/CD340 protein, rat (HEK293, His) | >95.0% | 100 UG
SDP

Catalog No. 5000293673
Encompass_Preferred

Recombinant rat HER2/CD340 extracellular domain produced in HEK293 cells and provided as a lyophilized preparation with a C-terminal 10xHis tag. Intended for research applications including biochemical assays, antibody binding studies, and cell signaling experiments. Purity is >95% by reducing SDS-PAGE; molecular weight approximately 91 kDa. Formulated in 20 mM PB, 150 mM NaCl, pH 7.4. Store lyophilized at -20°C and aliquot for long-term storage at -20°C or -80°C after reconstitution.

  • Recombinantly expressed in HEK293 cells.
  • C-terminal 10xHis tag for detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 91 kDa.
  • Lyophilized formulation in 20 mM PB, 150 mM NaCl, pH 7.4.
  • Suitable for binding assays, antibody characterization, and signaling studies.
  • Recommended storage: lyophilized at -20°C; aliquot and store at -20°C or -80°C after reconstitution.

Catalog No. 50-002-93673 Supplier Medchemexpress LLC Supplier No. HYP74895100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat HER2/CD340 extracellular domain produced in HEK293 cells and provided as a lyophilized preparation with a C-terminal 10xHis tag. Intended for research applications including biochemical assays, antibody binding studies, and cell signaling experiments. Purity is >95% by reducing SDS-PAGE; molecular weight approximately 91 kDa. Formulated in 20 mM PB, 150 mM NaCl, pH 7.4. Store lyophilized at -20°C and aliquot for long-term storage at -20°C or -80°C after reconstitution.

  • Recombinantly expressed in HEK293 cells.
  • C-terminal 10xHis tag for detection and purification.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 91 kDa.
  • Lyophilized formulation in 20 mM PB, 150 mM NaCl, pH 7.4.
  • Suitable for binding assays, antibody characterization, and signaling studies.
  • Recommended storage: lyophilized at -20°C; aliquot and store at -20°C or -80°C after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.