Learn More
Medchemexpress LLC Her2/CD340 protein, rat (HEK293, His) | >95.0% | 5 UG
SDP
Catalog No. 5000293688
Encompass_Preferred
Recombinant rat HER2/CD340 protein expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified ectodomain suitable for biochemical assays, antibody development, receptor binding studies, and structural or biophysical analysis. Molecular weight is approximately 91 kDa and purity is greater than 95% by reducing SDS-PAGE.
  • Recombinant rat HER2 protein produced in HEK293 expression system.
  • C-terminal His tag for efficient purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 91 kDa.
  • Suitable for biochemical assays, antibody generation, and binding studies.
  • Available in small research quantities (for example, 5 μg).
Catalog No. 50-002-93688 Supplier Medchemexpress LLC Supplier No. HYP748955UG
Recombinant rat HER2/CD340 protein expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified ectodomain suitable for biochemical assays, antibody development, receptor binding studies, and structural or biophysical analysis. Molecular weight is approximately 91 kDa and purity is greater than 95% by reducing SDS-PAGE.
  • Recombinant rat HER2 protein produced in HEK293 expression system.
  • C-terminal His tag for efficient purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight 91 kDa.
  • Suitable for biochemical assays, antibody generation, and binding studies.
  • Available in small research quantities (for example, 5 μg).