Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Her2/cd340 protein, rhesus macaque (HEK293, His) | >95.0% | 10 UG
SDP

Catalog No. 5000292461
Encompass_Preferred

Recombinant HER2/CD340 protein derived from rhesus macaque, expressed in HEK293 cells and supplied with a C-terminal His tag. Purified (>95% purity) and formulated for use in biochemical assays, antibody development, binding studies, and assay controls.

  • Recombinant extracellular domain of HER2 protein.
  • Derived from rhesus macaque.
  • Expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Available in microgram-scale quantities, including 10 μg.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week.
  • Suitable for biochemical assays, antibody development, and binding studies.

Catalog No. 50-002-92461 Supplier Medchemexpress LLC Supplier No. HYP7580710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant HER2/CD340 protein derived from rhesus macaque, expressed in HEK293 cells and supplied with a C-terminal His tag. Purified (>95% purity) and formulated for use in biochemical assays, antibody development, binding studies, and assay controls.

  • Recombinant extracellular domain of HER2 protein.
  • Derived from rhesus macaque.
  • Expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Available in microgram-scale quantities, including 10 μg.
  • Store at -20°C; after reconstitution stable at 4°C for 1 week.
  • Suitable for biochemical assays, antibody development, and binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.