Learn More
Carnabio Usa Inc HER2(ERBB2)/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 080165
Encompass
HER2(ERBB2), Cytoplasmic domain, Wild type, Amino Acid 676-1255, NP_004439.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0455