Learn More
Sino Biological Recombinant Anti-Her2/ERBB2 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 10004R205H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human HER2 / ERBB2 (rh HER2 / ERBB2; Catalog#10004-H08H; NP_004439.2; Met1-Thr652), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-7758