Learn More
Sino Biological Anti-Her2/ERBB2 Antibody (Biotin), Mouse Monoclonal
Supplier:  Sino Biological 10004MM01B100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ErbB2 / HER2 extracellular domain (rhErbB2; Catalog#10004-H08H; NP_004439.2; Met 1-Thr 652). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-228-3184