Learn More
Sino Biological HER2 / ErbB2 / CD340 Antibody (Biotin), Rabbit PAb 100ug
Supplier:  Sino Biological 10004RP03B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ErbB2 / HER2 extracellular domain (rhErbB2; Catalog#10004-H08H; NP_004439.2; Met 1-Thr652). Total IgG was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-224-6277