Learn More
Genscript Corporation Her2/ErbB2 His Mouse
Catalog No. 5000058312
Encompass
ErbB2 also called Neu and Her2 (human epidermal growth factor receptor 2) is a type I membrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors ErbB family members serve as receptors for the epidermal growth factor (EGF) family of growth factors Upon ERBB2 activation the MEMO1-RHOA-DIAPH1 signaling pathway elicits the phosphorylation and thus the inhibition of GSK3B at cell membrane This prevents the phosphorylation of APC and CLASP2 allowing its association with the cell membrane
Catalog No. 50-000-58312 Supplier Genscript Corporation Supplier No. Z03904500
Add to Cart
Add to Cart
ErbB2 also called Neu and Her2 (human epidermal growth factor receptor 2) is a type I membrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors ErbB family members serve as receptors for the epidermal growth factor (EGF) family of growth factors Upon ERBB2 activation the MEMO1-RHOA-DIAPH1 signaling pathway elicits the phosphorylation and thus the inhibition of GSK3B at cell membrane This prevents the phosphorylation of APC and CLASP2 allowing its association with the cell membrane