Learn More
Genscript Corporation Her2/ErbB2Biotin His Avi
Supplier:  Genscript Corporation Z03901100
Encompass
ErbB2 also called Neu and Her2 (human epidermal growth factor receptor 2) is a type I membrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors ErbB family members serve as receptors for the epidermal growth factor (EGF) family of growth factors Upon ERBB2 activation the MEMO1-RHOA-DIAPH1 signaling pathway elicits the phosphorylation and thus the inhibition of GSK3B at cell membrane
Catalog No. 50-000-57439