Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human Her2 / ErbB2 Protein, His Tag
Catalog No. 501956726
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human Her2, His Tag (HE2-HF224) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1). It is the FITC labeled form of Human Her2, His Tag (HE2-H5225).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.2 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Trastuzumab at 0.5 μg/mL (100 μL/well) can bind FITC-Labeled Human Her2, His Tag (Cat. No. HE2-HF224) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-195-6726 Supplier ACROBiosystems Supplier No. HE2HF224200UG
Recombinant Protein;200UG;FITC-Labeled Human Her2, His Tag (HE2-HF224) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1). It is the FITC labeled form of Human Her2, His Tag (HE2-H5225).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.2 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Trastuzumab at 0.5 μg/mL (100 μL/well) can bind FITC-Labeled Human Her2, His Tag (Cat. No. HE2-HF224) with a linear range of 0.02-0.313 μg/mL (QC tested).