Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus Her2 / ErbB2 Protein, His,Avitag™
Catalog No. 501956890
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Cynomolgus Her2, His,Avitag (HE2-C82E3) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # XP_005584091.2).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 72.9 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Trastuzumab at 3 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus Her2, His,Avitag (Cat. No. HE2-C82E3) with a linear range of 0.3-20 ng/mL (QC tested).
Catalog No. 50-195-6890 Supplier ACROBiosystems Supplier No. HE2C82E3200UG
Recombinant Protein;200UG;Biotinylated Cynomolgus Her2, His,Avitag (HE2-C82E3) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # XP_005584091.2).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 72.9 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Trastuzumab at 3 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus Her2, His,Avitag (Cat. No. HE2-C82E3) with a linear range of 0.3-20 ng/mL (QC tested).