Learn More
ACROBiosystems ACROBiosystems Mouse Her2 / ErbB2 Protein, His Tag
Supplier: ACROBiosystems ER2M5220100UG
Recombinant Protein;100UG;Mouse Her2, His Tag (ER2-M5220) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 653 (Accession # AAH53078.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.6 kDa. The protein migrates as 80-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.