Learn More
Sino Biological Recombinant Human EphB4 Protein (I782S, GST Tag)
Catalog No. 502575072
Encompass_Preferred
Recombinant human EPHB4 (I782S) (561-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5072 Supplier Sino Biological Supplier No. E2412G500
Recombinant human EPHB4 (I782S) (561-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.