Learn More
Medchemexpress LLC HER2/neu (654-662) GP2 | 160790-21-6 | 98.3% | 884.11 | 5 MG
SDP
Catalog No. 5000422770
Encompass_Preferred
HER2/neu (654-662) GP2 is a nine-amino acid peptide derived from the human epidermal growth factor receptor 2 (HER2/neu, 654-662). This peptide is known to induce HLA-A2-restricted cytotoxic T lymphocytes (CTL) that are reactive to various epithelial cancers. It is intended for research use only and not for patient use.
  • Derived from human epidermal growth factor receptor 2 (HER2/neu, 654-662).
  • Induces HLA-A2-restricted cytotoxic T lymphocytes (CTL).
  • CTLs are reactive to various epithelial cancers.
  • Solid, white to off-white appearance.
  • Sequence: Ile-Ile-Ser-Ala-Val-Val-Gly-Ile-Leu (IISAVVGIL).
Catalog No. 50-004-22770 Supplier Medchemexpress LLC Supplier No. HYP18555MG
HER2/neu (654-662) GP2 is a nine-amino acid peptide derived from the human epidermal growth factor receptor 2 (HER2/neu, 654-662). This peptide is known to induce HLA-A2-restricted cytotoxic T lymphocytes (CTL) that are reactive to various epithelial cancers. It is intended for research use only and not for patient use.
  • Derived from human epidermal growth factor receptor 2 (HER2/neu, 654-662).
  • Induces HLA-A2-restricted cytotoxic T lymphocytes (CTL).
  • CTLs are reactive to various epithelial cancers.
  • Solid, white to off-white appearance.
  • Sequence: Ile-Ile-Ser-Ala-Val-Val-Gly-Ile-Leu (IISAVVGIL).