Learn More
Diagnostic Biosystems HER2/NEU C-ERBB-2 c-erbB-2 Oncoprotein (SP3)
Catalog No. 502407330
Encompass_Preferred
This antibody recognizes a c-erbB-2 protein, which is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a subset of breast and other adenocarcinomas, as well astransitional cell carcinomas. In the case of breast cancer, expression determined byimmunohistochemistry has been shown to be associated with poor prognosis.
Catalog No. 50-240-7330 Supplier Diagnostic Biosystems Supplier No. RMPD008
This antibody recognizes a c-erbB-2 protein, which is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a subset of breast and other adenocarcinomas, as well astransitional cell carcinomas. In the case of breast cancer, expression determined byimmunohistochemistry has been shown to be associated with poor prognosis.