Learn More
Enzo Life Sciences HER2/Neu monoclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS3810100
Encompass_Preferred
Isotype: IgG. Immunogen: Recombinant HER2. Species reactivity: Human. Applications: IHC. Purity: Affinity-purified. Formulation: Liquid. In Tris buffer, pH 7.5, containing 1% BSA and <0.1% sodium azide.
Catalog No. 50-201-2908