Learn More
Enzo Life Sciences HER2 substrate (1mg)
SDP
Catalog No. 502011307
Encompass_Preferred
This peptide, GGMEDIYFEFMGGKKK, identified by screening a degenerate peptide library, is a substrate for several kinases, including HER-2/c-neu/erb-B-2, Tie-2, cFMS, and cKit. Alternative name: erbB-2 substrate. Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1307 Supplier Enzo Life Sciences Supplier No. BMLP2420001
This peptide, GGMEDIYFEFMGGKKK, identified by screening a degenerate peptide library, is a substrate for several kinases, including HER-2/c-neu/erb-B-2, Tie-2, cFMS, and cKit. Alternative name: erbB-2 substrate. Purity: ≥95%. Long Term Storage: -20°C.