Learn More
Enzo Life Sciences HER2 substrate (biotinylated) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2430001
Encompass_Preferred
This peptide, Biotin-Ahx-GGMEDIYFEFMGGKKK, identified by screening a degenerate peptide library, is a substrate for several kinases, including HER-2/c-neu/erb-B-2, Tie-2, cFMS, and cKit. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes. Alternative name: erbB-2 substrate (biotinylated). Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1308