Learn More
HER2 Trastuzumab297.rMAb anti-Human, BV605, Clone: Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb), BD OptiBuild™
Shop All Waters Corporation ProductsDescription
Specifications
Specifications
| Antigen | HER2 Trastuzumab297.rMAb |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb) |
| Concentration | 0.2 mg/mL |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | CD340; ERBB2; HER-2; HER-2/neu; HER2; NEU; proto-oncogene c-ErbB-2 |
| Host Species | Biosimilar |
| Immunogen | A431 Cell Line Expressing EGFR |
| Purification Method | Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.