Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (V842I, GST Tag), Active
Catalog No. 502575049
Encompass_Preferred
Recombinant human HER2 (V842I) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5049 Supplier Sino Biological Supplier No. E27122JG50
Recombinant human HER2 (V842I) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.