Learn More
Sino Biological Human HER2 / ErbB2 Protein (Fc Tag), Biotinylated 100ug
Supplier:  Sino Biological 10004H02HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Thr652) of human ErbB2 (NP_004439.2) was fused with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6162