Learn More
ACROBiosystems Recombinant Protein;Mouse Her2 / ErbB2 Protein;HEK293;1MG;ER2,ERBB2,CD340,HER-2,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1,ERBB-2,MLN-19,TKR-1
Catalog No. 501657724
Encompass_Preferred
Recombinant Protein;Mouse Her2 / ErbB2 Protein;1MG;Mouse Her2, His Tag (ER2-M5220) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 653 (Accession # AAH53078.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7724 Supplier ACROBiosystems Supplier No. ER2M52201MG
Recombinant Protein;Mouse Her2 / ErbB2 Protein;1MG;Mouse Her2, His Tag (ER2-M5220) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 653 (Accession # AAH53078.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.