Learn More
ACROBiosystems Recombinant Protein;Cynomolgus Her2 / ErbB2 Protein;HEK293;1MG;HE2,ERBB2,CD340,HER-2,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1,ERBB-2,MLN-19,TKR-1
Supplier:  ACROBiosystems HE2C52HB1MG
Encompass_Preferred
Recombinant Protein;Cynomolgus Her2 / ErbB2 Protein;1MG;Cynomolgus Her2 (HE2-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # XP_005584091.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 71.2 kDa. The protein migrates as 85-100 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Trastuzumab at 2 μg/mL (100 μL/well) can bind Cynomolgus Her2 (Cat. No. HE2-C52Hb) with a linear range of 2-6 ng/mL (QC tested).
Catalog No. 50-165-7685