Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human Her2 / ErbB2 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;HE2,ERBB2,CD340,HER-2,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1,ERBB-2,MLN-19,TKR-1
Catalog No. 501658011
Encompass_Preferred
Recombinant Protein;Biotinylated Human Her2 / ErbB2 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human Her2, His Tag, Avi Tag (HE2-H82E2) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 73.0 kDa. The protein migrates as 85-95 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Trastuzumab at 3 μg/mL (100 μL/well) can bind Biotinylated Human Her2, His Tag, Avi Tag (Cat. No. HE2-H82E2) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-165-8011 Supplier ACROBiosystems Supplier No. HE2H82E225UG
Recombinant Protein;Biotinylated Human Her2 / ErbB2 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human Her2, His Tag, Avi Tag (HE2-H82E2) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 73.0 kDa. The protein migrates as 85-95 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Trastuzumab at 3 μg/mL (100 μL/well) can bind Biotinylated Human Her2, His Tag, Avi Tag (Cat. No. HE2-H82E2) with a linear range of 0.2-6 ng/mL (QC tested).