Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (V956R, His Tag), Active
Catalog No. 502575038
Encompass_Preferred
Recombinant human HER2 (V956R) (677-1027) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-5038 Supplier Sino Biological Supplier No. E2712PH500
Recombinant human HER2 (V956R) (677-1027) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.