Learn More
Genscript Corporation Her3/ErbB3 hFc Chimera Human
Catalog No. 5000058845
Encompass
Her3 also called ErbB3 is a type I membrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors Her3 is expressed in keratinocytes melanocytes skeletal muscle cells embryonic myoblasts and Schwann cells Monomeric Her3 serves as a low affinity receptor for the heregulins (HRG)
Catalog No. 50-000-58845 Supplier Genscript Corporation Supplier No. Z03906500
Her3 also called ErbB3 is a type I membrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors Her3 is expressed in keratinocytes melanocytes skeletal muscle cells embryonic myoblasts and Schwann cells Monomeric Her3 serves as a low affinity receptor for the heregulins (HRG)