Learn More
Sino Biological HER3 / ErbB3 Neutralizing Antibody 500ug
Supplier:  Sino Biological 10201MH908500
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant Human HER3 / ErbB3. The antibody was produced using recombinant antibody technology.
Catalog No. 50-224-8256