Learn More
Genscript Corporation Her3 HisCynomolgus/Rhesus ma
Catalog No. 5000056714
Encompass
Her3 also called ErbB3 is a type I membrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors Her3 is expressed in keratinocytes melanocytes skeletal muscle cells embryonic myoblasts and Schwann cells Monomeric Her3 serves as a low affinity receptor for the heregulins (HRG)
Catalog No. 50-000-56714 Supplier Genscript Corporation Supplier No. Z03909100
Her3 also called ErbB3 is a type I membrane glycoprotein that is a member of the ErbB family of tyrosine kinase receptors Her3 is expressed in keratinocytes melanocytes skeletal muscle cells embryonic myoblasts and Schwann cells Monomeric Her3 serves as a low affinity receptor for the heregulins (HRG)