Learn More
Medchemexpress LLC HER3 Human HEK293 50ug
SDP
Catalog No. 5000181384
Encompass_Preferred
HER3 a pivotal tyrosine-protein kinase acts as a crucial cell receptor for neuregulins Neuregulin-1 (NRG1) activation boosts tyrosine phosphorylation and interaction with p85 subunit of phosphatidylinositol 3-kinase CSPG5 may also activate HER3 Its involvement in myeloid cell differentiation underscores HER3 s vital role in essential cellular processes for normal development and function HER3 Protein Human (HEK293 His solution) is the recombinant human-derived HER3 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-81384 Supplier Medchemexpress LLC Supplier No. HYP72625A50UG
HER3 a pivotal tyrosine-protein kinase acts as a crucial cell receptor for neuregulins Neuregulin-1 (NRG1) activation boosts tyrosine phosphorylation and interaction with p85 subunit of phosphatidylinositol 3-kinase CSPG5 may also activate HER3 Its involvement in myeloid cell differentiation underscores HER3 s vital role in essential cellular processes for normal development and function HER3 Protein Human (HEK293 His solution) is the recombinant human-derived HER3 protein expressed by HEK293 with C-6 His labeled tag