Learn More
Sino Biological Recombinant Human EphB4 Protein (His Tag), Active
Catalog No. 502575074
Encompass_Preferred
Recombinant human EPHB4 (561-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-5074 Supplier Sino Biological Supplier No. E2411H500
Recombinant human EPHB4 (561-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.