Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Her3 protein, rat (HEK293, His) | >95.0% | 1 MG
SDP

Catalog No. 5000291183
Encompass_Preferred

Recombinant rat HER3 protein expressed in HEK293 mammalian cells with a C-terminal His tag, supplied lyophilized for research applications in signaling and interaction studies. The protein is glycosylated, displays an apparent molecular weight of approximately 85-115 kDa, and is provided with analytical documentation for quality control.

  • Mammalian expression provides native glycosylation
  • C-terminal His tag for purification and detection
  • Apparent molecular weight ~85-115 kDa due to glycosylation
  • Purity >95.0% by reducing SDS-PAGE
  • Lyophilized from PBS (pH 7.4) for stability
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C long-term
  • Datasheet, COA, and SDS available for quality and safety information

Catalog No. 50-002-91183 Supplier Medchemexpress LLC Supplier No. HYP748911MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat HER3 protein expressed in HEK293 mammalian cells with a C-terminal His tag, supplied lyophilized for research applications in signaling and interaction studies. The protein is glycosylated, displays an apparent molecular weight of approximately 85-115 kDa, and is provided with analytical documentation for quality control.

  • Mammalian expression provides native glycosylation
  • C-terminal His tag for purification and detection
  • Apparent molecular weight ~85-115 kDa due to glycosylation
  • Purity >95.0% by reducing SDS-PAGE
  • Lyophilized from PBS (pH 7.4) for stability
  • Store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C long-term
  • Datasheet, COA, and SDS available for quality and safety information

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.