Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HER3 protein, rat (HEK293, His) | >95.0% | 50 UG
SDP

Catalog No. 5000293135
Encompass_Preferred

Recombinant rat HER3 (ErbB3) protein expressed in HEK293 cells, supplied as a C-terminal His-tagged, lyophilized powder. The product contains residues S20-H641, is glycosylated, and is provided as a 50 μg vial for biochemical and cell-based assays involving HER3 signaling and ligand interactions.

  • Expressed in HEK293 cells.
  • Contains a C-terminal His tag.
  • Includes residues S20-H641.
  • Lyophilized powder formulation from PBS, pH 7.4.
  • Purity >95.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg.
  • Apparent molecular weight 85-115 kDa due to glycosylation.
  • Recommend storage at -20°C; add carrier protein for long-term stability.

Catalog No. 50-002-93135 Supplier Medchemexpress LLC Supplier No. HYP7489150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat HER3 (ErbB3) protein expressed in HEK293 cells, supplied as a C-terminal His-tagged, lyophilized powder. The product contains residues S20-H641, is glycosylated, and is provided as a 50 μg vial for biochemical and cell-based assays involving HER3 signaling and ligand interactions.

  • Expressed in HEK293 cells.
  • Contains a C-terminal His tag.
  • Includes residues S20-H641.
  • Lyophilized powder formulation from PBS, pH 7.4.
  • Purity >95.0% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg.
  • Apparent molecular weight 85-115 kDa due to glycosylation.
  • Recommend storage at -20°C; add carrier protein for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.