Learn More
Cell Signaling Technology HER3/ErbB3 Monoclonal Antibody, Mouse Host, Unconjugated, 100 μg
SDP
Catalog No. 503132858
Encompass
The HER3/ErbB3 (E4D6O) Mouse Monoclonal Antibody (BSA and Azide Free) is a high-quality, carrier-free antibody designed for the detection of endogenous levels of total HER3/ErbB3 protein. Produced via immunization with a fragment of human ErbB3, this monoclonal antibody is validated for ELISA and ELISA-like assays.
  • Recognizes endogenous levels of total HER3/ErbB3 protein
  • Supplied in a BSA and Azide-free formulation, making it ideal for specialized or custom labeling with fluorophores, metals, lanthanides, and oligonucleotides
  • Quality control tested by size exclusion chromatography (SEC) to ensure antibody integrity
  • Validated for use in ELISA and ELISA-like assays
  • Specifically reactive to human ErbB3
Catalog No. 50-313-2858 Supplier Cell Signaling Technology Supplier No. 41239SF
The HER3/ErbB3 (E4D6O) Mouse Monoclonal Antibody (BSA and Azide Free) is a high-quality, carrier-free antibody designed for the detection of endogenous levels of total HER3/ErbB3 protein. Produced via immunization with a fragment of human ErbB3, this monoclonal antibody is validated for ELISA and ELISA-like assays.
  • Recognizes endogenous levels of total HER3/ErbB3 protein
  • Supplied in a BSA and Azide-free formulation, making it ideal for specialized or custom labeling with fluorophores, metals, lanthanides, and oligonucleotides
  • Quality control tested by size exclusion chromatography (SEC) to ensure antibody integrity
  • Validated for use in ELISA and ELISA-like assays
  • Specifically reactive to human ErbB3