Learn More
Medchemexpress LLC HER4 Human HEK293 50ug
SDP
Catalog No. 5000180471
Encompass_Preferred
HER4 is an important tyrosine protein kinase receptor for members of the neuregulin and EGF families that directs heart central nervous system and mammary gland development It is critical for myocardial differentiation postnatal cardiomyocyte proliferation neural crest cell migration axon guidance and mammary gland function HER4 Protein Human (HEK293 His-Fc) is the recombinant human-derived HER4 protein expressed by HEK293 with C-hFc C-10 His labeled tag
Catalog No. 50-001-80471 Supplier Medchemexpress LLC Supplier No. HYP7303550UG
HER4 is an important tyrosine protein kinase receptor for members of the neuregulin and EGF families that directs heart central nervous system and mammary gland development It is critical for myocardial differentiation postnatal cardiomyocyte proliferation neural crest cell migration axon guidance and mammary gland function HER4 Protein Human (HEK293 His-Fc) is the recombinant human-derived HER4 protein expressed by HEK293 with C-hFc C-10 His labeled tag